Assay Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007953.2 | NP_031979.2 | 2 - 3 | 522 | 61 | |
| GenBank mRNA | AK019222.1 | - | 2 - 3 | 483 | 61 | |
| AK156371.1 | - | 2 - 3 | 498 | 61 | ||
| BC034165.1 | - | 2 - 3 | 465 | 61 | ||
| BC039774.1 | - | 2 - 3 | 534 | 61 | ||
| BC138586.1 | - | 2 - 3 | 482 | 8861606 | 61 | |
| BC171958.1 | - | 2 - 3 | 482 | 9054244 | 61 | |
| CO425338.1 | - | 1 - 2 | 172 | 61 | ||
| U85259.1 | - | 2 - 3 | 489 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Esrra
Genomic Map