Assay Details
| Gene Symbol: Esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB560752.1 | - | 6 - 7 | 1457 | 63 | |
| AB560757.1 | - | 2 - 3 | 514 | 63 | ||
| AB560758.1 | - | 2 - 3 | 240 | 63 | ||
| AB560759.1 | - | 2 - 3 | 238 | 63 | ||
| AB560760.1 | - | 2 - 3 | 190 | 63 | ||
| AF128220.1 | - | 2 - 3 | 447 | 63 | ||
| AF128221.1 | - | 2 - 3 | 447 | 63 | ||
| AK036627.1 | - | 6 - 7 | 1431 | 63 | ||
| AK041525.1 | - | 7 - 8 | 1579 | 63 | ||
| AK077236.1 | - | 6 - 7 | 1437 | 63 | ||
| AK136228.1 | - | 2 - 3 | 236 | 63 | ||
| EU791538.1 | - | 6 - 7 | 1375 | 63 | ||
| EU791540.1 | - | 6 - 7 | 1500 | 63 | ||
| M38651.1 | - | 6 - 7 | 1439 | 63 | ||
More Information
Set Membership: |
|

Esr1
Genomic Map