Assay Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161373.1 | NP_001154845.1 | 5 - 6 | 1081 | 130 | |
| NM_001161374.1 | NP_001154846.1 | 5 - 6 | 1081 | 130 | ||
| NM_007977.2 | NP_032003.2 | 5 - 6 | 1081 | 130 | ||
| GenBank mRNA | AJ627040.1 | - | 4 - 5 | 501 | 130 | |
| AK051579.1 | - | 5 - 6 | 3357 | 130 | ||
| BC138347.1 | - | 5 - 6 | 962 | 8861367 | 130 | |
| BC145245.1 | - | 5 - 6 | 962 | 9053797 | 130 | |
| L05573.1 | - | 5 - 6 | 1079 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

F8
Genomic Map