Assay Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007977.2 | NP_032003.2 | 12 - 13 | 2310 | 74 | |
| GenBank mRNA | L05573.1 | - | 12 - 13 | 2308 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

F8
Genomic Map