Assay Details
| Gene Symbol: Fabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007980.3 | NP_032006.1 | 1 - 2 | 409 | 89 | |
| GenBank mRNA | AA245078.1 | - | 1 - 2 | 409 | 89 | |
| AK007995.1 | - | 1 - 2 | 107 | 89 | ||
| AY523818.1 | - | 1 - 2 | 95 | 89 | ||
| AY523819.1 | - | 1 - 2 | 95 | 89 | ||
| BC013457.1 | - | 1 - 2 | 89 | 4218050 | 89 | |
| BY707992.1 | - | 1 - 2 | 105 | 89 | ||
| M65034.1 | - | 1 - 2 | 103 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp2
Genomic Map