Assay Details
| Gene Symbol: Fgf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008005.2 | NP_032031.1 | 1 - 2 | 158 | 73 | |
| GenBank mRNA | AF075291.1 | - | 1 - 2 | 166 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF075291.1,NM_008005.2 transcripts.
Set Membership: |
|

Fgf18
Genomic Map