Assay Details
| Gene Symbol: Fgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008006.2 | NP_032032.1 | 1 - 2 | 370 | 61 | |
| GenBank mRNA | AF065903.1 | - | 1 - 2 | 173 | 61 | |
| AF065904.1 | - | 1 - 2 | 173 | 61 | ||
| AF065905.1 | - | 1 - 2 | 173 | 61 | ||
| AY027551.1 | - | 1 - 2 | 173 | 61 | ||
| BY730691.1 | - | 1 - 2 | 348 | 61 | ||
| M30644.1 | - | 1 - 2 | 173 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf2
Genomic Map