Assay Details
| Gene Symbol: Fgfr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163215.2 | NP_001156687.1 | 12 - 13 | 1965 | 75 | |
| NM_001163216.2 | NP_001156688.1 | 10 - 11 | 1680 | 75 | ||
| NM_001163217.2 | NP_001156689.1 | 11 - 12 | 1740 | 75 | ||
| NM_001205270.1 | NP_001192199.1 | 12 - 13 | 1942 | 75 | ||
| NM_008010.5 | NP_032036.2 | 12 - 13 | 1939 | 75 | ||
| GenBank mRNA | AF024638.1 | - | 10 - 11 | 1588 | 75 | |
| AK143592.1 | - | 12 - 13 | 1875 | 75 | ||
| AK163495.1 | - | 10 - 11 | 1518 | 75 | ||
| BC053056.1 | - | 12 - 13 | 1849 | 5708838 | 75 | |
| M81342.1 | - | 12 - 13 | 1857 | 75 | ||
| S56291.1 | - | 11 - 12 | 1684 | 75 | ||
| X58255.1 | - | 12 - 13 | 1843 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr3
Genomic Map