Assay Details
| Gene Symbol: Fgfr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163215.2 | NP_001156687.1 | 15 - 16 | 2347 | 150 | |
| NM_001163216.2 | NP_001156688.1 | 13 - 14 | 2062 | 150 | ||
| NM_001163217.2 | NP_001156689.1 | 14 - 15 | 2122 | 150 | ||
| NM_001205270.1 | NP_001192199.1 | 15 - 16 | 2324 | 150 | ||
| NM_008010.5 | NP_032036.2 | 15 - 16 | 2321 | 150 | ||
| GenBank mRNA | AF024638.1 | - | 13 - 14 | 1970 | 150 | |
| AK143592.1 | - | 15 - 16 | 2257 | 150 | ||
| AK163495.1 | - | 13 - 14 | 1900 | 150 | ||
| BC053056.1 | - | 15 - 16 | 2231 | 5708838 | 150 | |
| M81342.1 | - | 15 - 16 | 2239 | 153 | ||
| S56291.1 | - | 14 - 15 | 2066 | 150 | ||
| X58255.1 | - | 15 - 16 | 2225 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr3
Genomic Map