Assay Details
| Gene Symbol: Fyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122892.1 | NP_001116364.1 | 9 - 10 | 1474 | 78 | |
| NM_001122893.1 | NP_001116365.1 | 10 - 11 | 1553 | 78 | ||
| NM_008054.2 | NP_032080.2 | 10 - 11 | 1544 | 78 | ||
| GenBank mRNA | AK156584.1 | - | 10 - 11 | 1555 | 78 | |
| AK171646.1 | - | 10 - 11 | 1546 | 78 | ||
| BC010554.1 | - | 9 - 10 | 1155 | 78 | ||
| BC032149.1 | - | 9 - 10 | 1189 | 3602546 | 78 | |
| BC092217.1 | - | 9 - 10 | 1470 | 6403711 | 78 | |
| M27266.1 | - | 11 - 12 | 1256 | 78 | ||
| U70324.1 | - | 10 - 11 | 1257 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fyn
Genomic Map