Assay Details
| Gene Symbol: Fzd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008055.4 | NP_032081.3 | 1 - 2 | 603 | 70 | |
| GenBank mRNA | AK044794.1 | - | 1 - 2 | 593 | 70 | |
| AK049945.1 | - | 1 - 2 | 517 | 70 | ||
| AK050891.1 | - | 1 - 2 | 576 | 70 | ||
| AK132577.1 | - | 1 - 2 | 586 | 70 | ||
| BC015256.1 | - | 1 - 2 | 603 | 4238940 | 70 | |
| U43317.1 | - | 1 - 2 | 609 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK049945.1 transcripts.
Set Membership: |
|

Fzd4
Genomic Map