Assay Details
| Gene Symbol: Fzd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162494.1 | NP_001155966.1 | 5 - 6 | 1858 | 85 | |
| NM_008056.3 | NP_032082.3 | 5 - 6 | 1862 | 85 | ||
| GenBank mRNA | AK088399.1 | - | 5 - 6 | 1727 | 85 | |
| AK138964.1 | - | 2 - 3 | 575 | 85 | ||
| BC026150.1 | - | 5 - 6 | 1851 | 3983985 | 85 | |
| U43319.1 | - | 4 - 5 | 1688 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fzd6
Genomic Map