| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100003677.mm.1 |
NM_008057.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003677.mm.1 |
NM_008057.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003677.mm.1 |
TC0100000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100003679.mm.1 |
NM_008057.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003679.mm.1 |
NM_008057.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003679.mm.1 |
TC0100000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100003675.mm.1 |
NM_008057.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003675.mm.1 |
NM_008057.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003675.mm.1 |
TC0100000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100003676.mm.1 |
NM_008057.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003676.mm.1 |
NM_008057.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003676.mm.1 |
TC0100000450.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100003678.mm.1 |
NM_008057.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003678.mm.1 |
NM_008057.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003678.mm.1 |
TC0100000450.mm.1 |
MTA TC |