Assay Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008067.4 | NP_032093.3 | 2 - 3 | 419 | 132 | |
| GenBank mRNA | AK039144.1 | - | 2 - 3 | 360 | 132 | |
| AK134760.1 | - | 2 - 3 | 328 | 132 | ||
| BC119317.1 | - | 2 - 3 | 217 | 8734066 | 132 | |
| BC120686.1 | - | 2 - 3 | 217 | 40129609 | 132 | |
| M86568.1 | - | 2 - 3 | 321 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabra3
Genomic Map