Assay Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008067.4 | NP_032093.3 | 5 - 6 | 829 | 111 | |
| GenBank mRNA | AK039144.1 | - | 5 - 6 | 770 | 111 | |
| BC119317.1 | - | 5 - 6 | 627 | 8734066 | 111 | |
| BC120686.1 | - | 5 - 6 | 627 | 40129609 | 111 | |
| M86568.1 | - | 5 - 6 | 731 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabra3
Genomic Map