Assay Details
| Gene Symbol: Gabrr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008076.3 | NP_032102.2 | 4 - 5 | 649 | 70 | |
| GenBank mRNA | AF024621.1 | - | 4 - 5 | 762 | 70 | |
| AK132642.1 | - | 4 - 5 | 477 | 70 | ||
| AK160446.1 | - | 2 - 3 | 316 | 70 | ||
| BC057957.1 | - | 4 - 5 | 634 | 5357713 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrr2
Genomic Map