Assay Details
| Gene Symbol: Opn1mw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008106.2 | NP_032132.1 | 2 - 3 | 434 | 84 | |
| GenBank mRNA | AF011389.1 | - | 2 - 3 | 420 | 84 | |
| AF190672.1 | - | 2 - 3 | 436 | 84 | ||
| AK149287.1 | - | 2 - 3 | 435 | 84 | ||
| BC014826.1 | - | 2 - 3 | 414 | 4507311 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Opn1mw
Genomic Map