Assay Details
| Gene Symbol: Gdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008108.4 | NP_032134.2 | 1 - 2 | 373 | 70 | |
| GenBank mRNA | AK082877.1 | - | 1 - 2 | 294 | 70 | |
| AK146174.1 | - | 1 - 2 | 299 | 70 | ||
| AK152457.1 | - | 1 - 2 | 374 | 70 | ||
| AK160533.1 | - | 1 - 2 | 331 | 70 | ||
| AK167962.1 | - | 1 - 2 | 299 | 70 | ||
| BC101963.1 | - | 1 - 2 | 288 | 40045099 | 70 | |
| BC101964.1 | - | 1 - 2 | 288 | 40045100 | 70 | |
| BC101965.1 | - | 1 - 2 | 71 | |||
| BC103565.1 | - | 1 - 2 | 287 | 40045101 | 70 | |
| L06443.1 | - | 1 - 2 | 389 | 70 | ||
| S52658.1 | - | 1 - 2 | 329 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdf3
Genomic Map