Assay Details
| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008118.3 | NP_032144.2 | 7 - 8 | 1156 | 93 | |
| GenBank mRNA | AK078933.1 | - | 7 - 8 | 1157 | 93 | |
| BC118519.1 | - | 7 - 8 | 1142 | 40105765 | 93 | |
| L24191.1 | - | 7 - 8 | 1068 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gif
Genomic Map