Assay Details
| Gene Symbol: Gpr65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008152.3 | NP_032178.1 | 1 - 2 | 76 | 72 | |
| GenBank mRNA | AK134113.1 | - | 1 - 2 | 77 | 72 | |
| AK142853.1 | - | 1 - 2 | 40 | 72 | ||
| U39827.1 | - | 1 - 2 | 54 | 72 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK134113.1,AK142853.1,NM_008152.3,U39827.1 transcripts.
Set Membership: |
|

Gpr65
Genomic Map