Assay Details
| Gene Symbol: Grik5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008168.2 | NP_032194.2 | 4 - 5 | 723 | 92 | |
| GenBank mRNA | BC052009.1 | - | 4 - 5 | 611 | 5695215 | 92 |
| BC058097.1 | - | 4 - 5 | 482 | 92 | ||
| BC110682.1 | - | 4 - 5 | 646 | 5033584 | 92 | |
| D10011.1 | - | 4 - 5 | 652 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik5
Genomic Map