Assay Details
| Gene Symbol: Grik5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008168.2 | NP_032194.2 | 8 - 9 | 1287 | 62 | |
| GenBank mRNA | BC052009.1 | - | 8 - 9 | 1175 | 5695215 | 62 |
| BC058097.1 | - | 8 - 9 | 1046 | 62 | ||
| BC110682.1 | - | 8 - 9 | 1210 | 5033584 | 62 | |
| D10011.1 | - | 8 - 9 | 1216 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik5
Genomic Map