Assay Details
| Gene Symbol: Grik5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008168.2 | NP_032194.2 | 16 - 17 | 2413 | 56 | |
| GenBank mRNA | BC052009.1 | - | 16 - 17 | 2301 | 5695215 | 56 |
| BC058097.1 | - | 16 - 17 | 2172 | 56 | ||
| BC110682.1 | - | 16 - 17 | 2336 | 5033584 | 56 | |
| D10011.1 | - | 16 - 17 | 2342 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grik5
Genomic Map