Assay Details
| Gene Symbol: Grin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008169.3 | NP_032195.1 | 18 - 19 | 2891 | 57 | |
| GenBank mRNA | AW494928.1 | - | 2 - 3 | 57 | ||
| D10028.1 | - | 18 - 19 | 2683 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grin1
Genomic Map