Assay Details
| Gene Symbol: Grin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008169.3 | NP_032195.1 | 19 - 20 | 3000 | 90 | |
| GenBank mRNA | D10028.1 | - | 19 - 20 | 2792 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grin1
Genomic Map