Assay Details
| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008171.3 | NP_032197.3 | 4 - 5 | 1180 | 114 | |
| GenBank mRNA | AK002963.1 | - | 4 - 5 | 1181 | 114 | |
| D10651.2 | - | 1 - 2 | 417 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grin2b
Genomic Map