Assay Details
| Gene Symbol: Grpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033473.1 | - | 1 - 2 | 779 | 118 | |
| BC113145.1 | - | 1 - 2 | 667 | 40047100 | 118 | |
| M57922.1 | - | 1 - 2 | 795 | 118 | ||
| M61000.1 | - | 1 - 2 | 768 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grpr
Genomic Map