Assay Details
| Gene Symbol: Gyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294138.1 | NP_001281067.1 | 12 - 13 | 1275 | 65 | |
| NM_001294140.1 | NP_001281069.1 | 12 - 13 | 1209 | 65 | ||
| NM_008194.3 | NP_032220.1 | 11 - 12 | 1191 | 65 | ||
| NM_212444.2 | NP_997609.1 | 11 - 12 | 1191 | 65 | ||
| GenBank mRNA | AK044308.1 | - | 12 - 13 | 1047 | 65 | |
| AK088373.1 | - | 11 - 12 | 1010 | 65 | ||
| AK169424.1 | - | 12 - 13 | 1081 | 65 | ||
| AK169525.1 | - | 11 - 12 | 1192 | 65 | ||
| BC003767.1 | - | 11 - 12 | 994 | 3593896 | 65 | |
| BQ555961.1 | - | 1 - 2 | 56 | 65 | ||
| CB950214.1 | - | 6 - 7 | 373 | 65 | ||
| CK789590.1 | - | 6 - 7 | 341 | 65 | ||
| U48403.1 | - | 11 - 12 | 968 | 65 | ||
More Information
Set Membership: |
|

Gyk
Genomic Map