Assay Details
| Gene Symbol: Slc39a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077709.1 | NP_001071177.1 | 5 - 6 | 965 | 67 | |
| NM_008202.2 | NP_032228.2 | 4 - 5 | 1250 | 67 | ||
| GenBank mRNA | AK087839.1 | - | 4 - 5 | 945 | 67 | |
| AK145956.1 | - | 3 - 4 | 1215 | 67 | ||
| AK159968.1 | - | 4 - 5 | 1126 | 67 | ||
| AK167344.1 | - | 3 - 4 | 1298 | 67 | ||
| BC115426.2 | - | 4 - 5 | 1104 | 40093613 | 67 | |
| BC115427.1 | - | 4 - 5 | 1104 | 40093614 | 67 | |
| KJ650218.1 | - | 4 - 5 | 818 | 67 | ||
| M32010.1 | - | 4 - 5 | 1118 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a7
Genomic Map