Assay Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008261.2 | NP_032287.2 | 2 - 3 | 404 | 79 | |
| GenBank mRNA | AF015275.1 | - | 2 - 3 | 326 | 79 | |
| AK143948.1 | - | 2 - 3 | 429 | 79 | ||
| BC039220.1 | - | 2 - 3 | 404 | 4238842 | 79 | |
| D29015.1 | - | 2 - 3 | 331 | 79 | ||
| EF193391.1 | - | 2 - 3 | 203 | 79 | ||
| EF193393.1 | - | 2 - 3 | 229 | 79 | ||
| FJ608819.1 | - | 3 - 4 | 358 | 79 | ||
| FJ608820.1 | - | 3 - 4 | 358 | 79 | ||
| FJ608821.1 | - | 3 - 4 | 358 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnf4a
Genomic Map