Assay Details
| Gene Symbol: Lipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008280.2 | NP_032306.2 | 1 - 2 | 336 | 89 | |
| GenBank mRNA | AY228765.1 | - | 1 - 2 | 88 | 89 | |
| BC021841.1 | - | 1 - 2 | 336 | 5124139 | 89 | |
| BC094050.1 | - | 1 - 2 | 123 | 5135403 | 89 | |
| X58426.1 | - | 1 - 2 | 107 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lipc
Genomic Map