Assay Details
| Gene Symbol: Lipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008280.2 | NP_032306.2 | 2 - 3 | 527 | 81 | |
| GenBank mRNA | AK158285.1 | - | 2 - 3 | 378 | 81 | |
| AK158360.1 | - | 2 - 3 | 378 | 81 | ||
| AY228765.1 | - | 2 - 3 | 279 | 81 | ||
| BC021841.1 | - | 2 - 3 | 527 | 5124139 | 81 | |
| BC094050.1 | - | 2 - 3 | 314 | 5135403 | 81 | |
| X58426.1 | - | 2 - 3 | 298 | 81 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK158285.1,AK158360.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
The orphan nuclear receptor, RORalpha, regulates gene expression that controls lipid metabolism: staggerer (SG/SG) mice are resistant to diet-induced obesity.
- Authors
- Lau P, Fitzsimmons RL, Raichur S, Wang SC, Lechtken A, Muscat GE
- Journal
- J Biol Chem - Page No 18411 (Fri Jun 27 00:00:00 EDT 2008)
- Volume (Year)
- 283 (2008)
- Product Usage
- -

Lipc
Genomic Map