Assay Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008280.2 | NP_032306.2 | 4 - 5 | 824 | 91 | |
GenBank mRNA | AK158285.1 | - | 4 - 5 | 675 | 91 | |
AK158360.1 | - | 4 - 5 | 675 | 91 | ||
AY228765.1 | - | 4 - 5 | 576 | 91 | ||
BC021841.1 | - | 4 - 5 | 824 | 5124139 | 91 | |
BC094050.1 | - | 4 - 5 | 611 | 5135403 | 91 | |
X58426.1 | - | 4 - 5 | 595 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
