Assay Details
| Gene Symbol: Lipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008280.2 | NP_032306.2 | 7 - 8 | 1417 | 101 | |
| GenBank mRNA | AK158285.1 | - | 7 - 8 | 1268 | 101 | |
| AK158360.1 | - | 7 - 8 | 1268 | 101 | ||
| AY228765.1 | - | 7 - 8 | 1169 | 101 | ||
| BC021841.1 | - | 7 - 8 | 1417 | 5124139 | 101 | |
| BC094050.1 | - | 7 - 8 | 1204 | 5135403 | 101 | |
| X58426.1 | - | 7 - 8 | 1188 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lipc
Genomic Map