Assay Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008339.2 | NP_032365.1 | 2 - 3 | 159 | 74 | |
| GenBank mRNA | AK143573.1 | - | 2 - 3 | 160 | 74 | |
| BC012226.1 | - | 2 - 3 | 159 | 4164551 | 74 | |
| CT010358.1 | - | 2 - 3 | 114 | 74 | ||
| J03857.1 | - | 2 - 3 | 298 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | J03857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | J03857.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492206 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172624 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115417 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168336 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | J03857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | J03857.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | J03857.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | J03857.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100880 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172624 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168336 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115417 | - | - |
| Whole Mouse Genome G4122A | A_51_P342652 | J03857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | J03857.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | BC012226.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | NM_008339.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342652 | AK143573.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140498 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110475 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172624 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168336 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115417 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09630544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09630542 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09510068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09611262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09611260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09402875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09402868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09402861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09611252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09402848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09510068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09402858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09402874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10392147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392145 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417640_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10392142 | - | - |
| Mouse Genome 430 2.0 Array | 1417640_at | - | - |
| Mouse Genome 430 2.0 Array | 1417640_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15985_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15985_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100036724.mm.1 | NM_008339.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100036724.mm.1 | TC1100003920.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417640_PM_at | - | - |
| Mouse Expression Set 430 | 1417640_at | - | - |
| Murine Genome U74v2 Set | 161012_PM_at | - | - |
| Murine Genome U74v2 Set | 161012_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417640_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417640_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417640_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417640_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 498083 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450066 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3390358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450066 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2661523 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2661523 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610822 | - | - |
| mouse-6 v1.1 (Illumina) | scl39449.6.1_88-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001391.1_166-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390358 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450066 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001391.1_166-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39449.6.1_88-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450066 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2661523 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2610822 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2610822 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2661523 | - | - |

Cd79b
Genomic Map