Assay Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008339.2 | NP_032365.1 | 3 - 4 | 475 | 61 | |
| GenBank mRNA | AK143573.1 | - | 3 - 4 | 476 | 61 | |
| BC012226.1 | - | 3 - 4 | 475 | 4164551 | 61 | |
| CT010358.1 | - | 3 - 4 | 430 | 61 | ||
| J03857.1 | - | 3 - 4 | 614 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd79b
Genomic Map