Assay Details
| Gene Symbol: Il12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF128210.1 | - | 2 - 3 | 173 | 68 | |
| AF128211.1 | - | 2 - 3 | 173 | 68 | ||
| AF128212.1 | - | 2 - 3 | 173 | 68 | ||
| AF128213.1 | - | 2 - 3 | 173 | 68 | ||
| BC116851.1 | - | 2 - 3 | 177 | 68 | ||
| BC116855.1 | - | 2 - 3 | 177 | 68 | ||
| M86672.1 | - | 2 - 3 | 275 | 68 | ||
More Information
Set Membership: |
|
Citations (2)
CCR7 ligands are required for development of experimental autoimmune encephalomyelitis through generating IL-23-dependent Th17 cells.
- Authors
- Kuwabara T, Ishikawa F, Yasuda T, Aritomi K, Nakano H, Tanaka Y, Okada Y, Lipp M, Kakiuchi T
- Journal
- J Immunol - Page No 2513 (Sat Aug 15 00:00:00 EDT 2009)
- Volume (Year)
- 183 (2009)
- Product Usage
- -
Role of IFN-gamma and IL-6 in a protective immune response to Yersinia enterocolitica in mice.
- Authors
- Matteoli G, Fahl E, Warnke P, Muller S, Bonin M, Autenrieth IB, Bohn E
- Journal
- Bmc Microbiol - Page No 153 (Tue Jan 01 00:00:00 EST 2008)
- Volume (Year)
- 8 (2008)
- Product Usage
- -

Il12a
Genomic Map