Assay Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008353.2 | NP_032379.2 | 10 - 11 | 1356 | 90 | |
| GenBank mRNA | AK137577.1 | - | 10 - 11 | 1357 | 90 | |
| AK171937.1 | - | 10 - 11 | 1333 | 90 | ||
| BC057212.1 | - | 10 - 11 | 1339 | 5350447 | 90 | |
| U23922.1 | - | 10 - 11 | 1311 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il12rb1
Genomic Map