Assay Details
| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254747.1 | NP_001241676.1 | 3 - 4 | 507 | 78 | |
| NM_008357.2 | NP_032383.1 | 3 - 4 | 517 | 78 | ||
| GenBank mRNA | AK155616.1 | - | 3 - 4 | 1155 | 78 | |
| BC023698.1 | - | 3 - 4 | 502 | 5345382 | 78 | |
| CJ137432.1 | - | 2 - 3 | 78 | |||
| U14332.1 | - | 3 - 4 | 476 | 78 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK155616.1,BC023698.1,NM_001254747.1,NM_008357.2,U14332.1 transcripts.
Set Membership: |
|

Il15
Genomic Map