Assay Details
| Gene Symbol: Il1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008361.3 | NP_032387.1 | 2 - 3 | 114 | 70 | |
| GenBank mRNA | AK168047.1 | - | 2 - 3 | 134 | 70 | |
| AK225002.1 | - | 2 - 3 | 133 | 70 | ||
| BC011437.1 | - | 2 - 3 | 114 | 3989461 | 70 | |
| M15131.1 | - | 2 - 3 | 123 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1b
Genomic Map