Assay Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008372.4 | NP_032398.3 | 3 - 4 | 515 | 114 | |
| GenBank mRNA | AF078906.1 | - | 3 - 4 | 381 | 114 | |
| AK040364.1 | - | 3 - 4 | 498 | 114 | ||
| AK040740.1 | - | 3 - 4 | 498 | 114 | ||
| AK041838.1 | - | 3 - 4 | 406 | 114 | ||
| AK042264.1 | - | 3 - 4 | 460 | 114 | ||
| AK154215.1 | - | 2 - 3 | 329 | 114 | ||
| AK154516.1 | - | 3 - 4 | 475 | 114 | ||
| BC089571.1 | - | 3 - 4 | 467 | 6758487 | 114 | |
| M29697.1 | - | 3 - 4 | 428 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il7r
Genomic Map