Assay Details
| Gene Symbol: Il7r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008372.4 | NP_032398.3 | 5 - 6 | 841 | 76 | |
| GenBank mRNA | AF078906.1 | - | 5 - 6 | 707 | 76 | |
| AK040364.1 | - | 5 - 6 | 824 | 76 | ||
| AK042264.1 | - | 5 - 6 | 786 | 76 | ||
| AK154215.1 | - | 4 - 5 | 655 | 76 | ||
| AK154516.1 | - | 5 - 6 | 801 | 76 | ||
| BC089571.1 | - | 5 - 6 | 793 | 6758487 | 76 | |
| M29697.1 | - | 5 - 6 | 754 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il7r
Genomic Map