Assay Details
| Gene Symbol: Itgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008404.4 | NP_032430.2 | 2 - 3 | 175 | 63 | |
| GenBank mRNA | AK088435.1 | - | 2 - 3 | 164 | 63 | |
| AK136502.1 | - | 2 - 3 | 176 | 63 | ||
| AK161236.1 | - | 2 - 3 | 144 | 63 | ||
| AK169587.1 | - | 2 - 3 | 150 | 63 | ||
| BC145644.1 | - | 1 - 2 | 97 | 40130947 | 63 | |
| M31039.1 | - | 1 - 2 | 63 | 63 | ||
| X14951.1 | - | 2 - 3 | 141 | 63 | ||
More Information
Set Membership: |
|

Itgb2
Genomic Map