Assay Details
| Gene Symbol: Itgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008404.4 | NP_032430.2 | 5 - 6 | 612 | 59 | |
| GenBank mRNA | AK088435.1 | - | 5 - 6 | 601 | 59 | |
| AK136502.1 | - | 5 - 6 | 613 | 59 | ||
| AK161236.1 | - | 5 - 6 | 581 | 59 | ||
| AK169587.1 | - | 5 - 6 | 647 | 59 | ||
| BC145644.1 | - | 4 - 5 | 534 | 40130947 | 59 | |
| M31039.1 | - | 4 - 5 | 500 | 59 | ||
| X14951.1 | - | 5 - 6 | 578 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb2
Genomic Map