Assay Details
| Gene Symbol: Itgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008404.4 | NP_032430.2 | 14 - 15 | 2196 | 75 | |
| GenBank mRNA | AK088435.1 | - | 14 - 15 | 2185 | 75 | |
| AK136502.1 | - | 14 - 15 | 2197 | 75 | ||
| AK161236.1 | - | 13 - 14 | 1920 | 75 | ||
| AK169587.1 | - | 12 - 13 | 1798 | 75 | ||
| BC145644.1 | - | 13 - 14 | 2118 | 40130947 | 75 | |
| M31039.1 | - | 13 - 14 | 2084 | 75 | ||
| X14951.1 | - | 14 - 15 | 2162 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itgb2
Genomic Map