Assay Details
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048177.2 | NP_001041642.1 | 22 - 23 | 3523 | 101 | |
| NM_008413.3 | NP_032439.2 | 23 - 24 | 3606 | 101 | ||
| GenBank mRNA | BC054807.1 | - | 22 - 23 | 3286 | 6838318 | 101 |
| BC059834.1 | - | 22 - 23 | 3286 | 6838318 | 101 | |
| L16956.1 | - | 22 - 23 | 3275 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jak2
Genomic Map