Assay Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008434.2 | NP_032460.2 | 1 - 2 | 486 | 62 | |
| GenBank mRNA | AB079603.1 | - | 1 - 2 | 189 | 62 | |
| AK154244.1 | - | 1 - 2 | 460 | 62 | ||
| AY331142.1 | - | 1 - 2 | 401 | 62 | ||
| BC045142.1 | - | 1 - 2 | 486 | 6314642 | 62 | |
| BC055304.1 | - | 1 - 2 | 441 | 6397978 | 62 | |
| U70068.1 | - | 1 - 2 | 292 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnq1
Genomic Map