Assay Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008434.2 | NP_032460.2 | 13 - 14 | 1789 | 66 | |
| GenBank mRNA | AK136389.1 | - | 6 - 7 | 659 | 66 | |
| AK154244.1 | - | 13 - 14 | 1763 | 66 | ||
| AY331142.1 | - | 13 - 14 | 1704 | 66 | ||
| BC045142.1 | - | 13 - 14 | 1789 | 6314642 | 66 | |
| BC055304.1 | - | 13 - 14 | 1744 | 6397978 | 66 | |
| U70068.1 | - | 13 - 14 | 1595 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnq1
Genomic Map