Assay Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008465.5 | NP_032491.2 | 5 - 6 | 586 | 93 | |
| GenBank mRNA | AK028259.1 | - | 5 - 6 | 587 | 93 | |
| AK028307.1 | - | 5 - 6 | 605 | 93 | ||
| AK154800.1 | - | 5 - 6 | 604 | 93 | ||
| AK169311.1 | - | 5 - 6 | 609 | 93 | ||
| AK169348.1 | - | 5 - 6 | 609 | 93 | ||
| AK218219.1 | - | 4 - 5 | 346 | 93 | ||
| BC006771.1 | - | 5 - 6 | 571 | 3588534 | 93 | |
| U20619.1 | - | 5 - 6 | 591 | 93 | ||
| U34228.1 | - | 4 - 5 | 435 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kpna1
Genomic Map