Assay Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008465.5 | NP_032491.2 | 9 - 10 | 1067 | 69 | |
| GenBank mRNA | AK028259.1 | - | 9 - 10 | 1068 | 69 | |
| AK028307.1 | - | 9 - 10 | 1086 | 69 | ||
| AK154800.1 | - | 9 - 10 | 1085 | 69 | ||
| AK169311.1 | - | 9 - 10 | 1090 | 69 | ||
| AK169348.1 | - | 9 - 10 | 1090 | 69 | ||
| BC006771.1 | - | 9 - 10 | 1052 | 3588534 | 69 | |
| U20619.1 | - | 9 - 10 | 1072 | 69 | ||
| U34228.1 | - | 8 - 9 | 916 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kpna1
Genomic Map